Learn More
Med Vet International IV3000™ Catheter Securement Dressing, 4 x 5 Inch, Sterile, 4925, 10/Box
Catalog No. 503043402
Encompass_Preferred
IV3000 prevents moisture from building up – this, in turn, reduces risk of infection The higher permeability of IV3000 keeps the skin drier so improves dressing adherence and reduces the number of unscheduled dressing changes IV3000 is transparent and so allows regular inspection of the catheter insertion site for signs of infection or phlebitis without the need for dressing removal IV3000 is designed to provide easy aseptic application and removal Film acts as a barrier to bacteria including MSRA 10 sterile transparent catheter securement dressings per box
Catalog No. 50-304-3402 Supplier Med Vet International Supplier No. 193313BX
IV3000 prevents moisture from building up – this, in turn, reduces risk of infection The higher permeability of IV3000 keeps the skin drier so improves dressing adherence and reduces the number of unscheduled dressing changes IV3000 is transparent and so allows regular inspection of the catheter insertion site for signs of infection or phlebitis without the need for dressing removal IV3000 is designed to provide easy aseptic application and removal Film acts as a barrier to bacteria including MSRA 10 sterile transparent catheter securement dressings per box