Learn More
Sklar Instruments Dressing Jar, Sklar, Stainless Steel. This jar measures 5-7/8 in. x 6-3/4 in. and has a capacity of 3 quarts.
SDP
Catalog No. 501437030 Shop All Sklar Instruments Products
Encompass_Preferred
Dressing Jar, Sklar, Stainless Steel. Sklar Dressing Jars are ideal for storing medical supplies such as tongue depressors, cotton balls, and cotton swabs. This jar measures 5-7/8 in. x 6-3/4 in. and has a capacity of 3 quarts. It also features a slip over lid and a recessed handle. This product is sold in packs of 12.
This item is not returnable. View return policy
Catalog No. 50-143-7030 Supplier Sklar Instruments Supplier No. 10152712
Add to Cart
Add to Cart
Dressing Jar, Sklar, Stainless Steel. Sklar Dressing Jars are ideal for storing medical supplies such as tongue depressors, cotton balls, and cotton swabs. This jar measures 5-7/8 in. x 6-3/4 in. and has a capacity of 3 quarts. It also features a slip over lid and a recessed handle. This product is sold in packs of 12.