Learn More
Med Vet International Foam Dressing Kendall™ 4 X 4 Inch Without Border Without Film Backing Nonadhesive Square Sterile, 55544, 10/Box
Catalog No. 503044468
Encompass_Preferred
Ultra-soft hydrophilic foam that protects and cushions wounds Small pores decrease the chance of tissue migration into the foam dressing Polyurethane top-sheet prevents strike-through while allowing for moisture-vapor transmission and helping to reduce skin maceration 4 x 4 inches
Catalog No. 50-304-4468 Supplier Med Vet International Supplier No. 543990BX
Ultra-soft hydrophilic foam that protects and cushions wounds Small pores decrease the chance of tissue migration into the foam dressing Polyurethane top-sheet prevents strike-through while allowing for moisture-vapor transmission and helping to reduce skin maceration 4 x 4 inches