Learn More
Med Vet International MedFix™ Nonwoven Dressing Retention Tape, 2 Inch x 11 Yard, White, MSC4002, 1/Box
Catalog No. 503043713
Encompass_Preferred
For flexibility and customized sizing Low sensitivity adhesive and therefore gentle Medfix has a printed s-c release liner Water resistant May be left in place for up to 7 days
Catalog No. 50-304-3713 Supplier Med Vet International Supplier No. 684035BX
For flexibility and customized sizing Low sensitivity adhesive and therefore gentle Medfix has a printed s-c release liner Water resistant May be left in place for up to 7 days