Learn More
Med Vet International Foam Dressing Mepilex 4 X 4 Inch Without Border Film Backing Silicone Adhesive Square Sterile, 294199, Each
Catalog No. 503044314
Encompass_Preferred
Mepilex is a soft and highly conformable foam dressing that absorbs exudate and maintains a moist wound environment The Safetac Technology layer seals the wound edges, preventing the exudate from leaking onto the surrounding skin, thus minimizing the risk of maceration Well suited for use under compression bandages May be cut to suit various wound shapes and difficult-to-dress locations Polyurethane backing film is breathable, moisture-proof and a bacterial and viral barrier
Catalog No. 50-304-4314 Supplier Med Vet International Supplier No. 712209EA
Mepilex is a soft and highly conformable foam dressing that absorbs exudate and maintains a moist wound environment The Safetac Technology layer seals the wound edges, preventing the exudate from leaking onto the surrounding skin, thus minimizing the risk of maceration Well suited for use under compression bandages May be cut to suit various wound shapes and difficult-to-dress locations Polyurethane backing film is breathable, moisture-proof and a bacterial and viral barrier