Learn More
Med Vet International Sodium Chloride Impregnated Dressing Mesalt Square 4 X 4 Inch / 2 X 2 Inch Folded Sterile, 285580, 30/Box
Catalog No. 503045469
Encompass_Preferred
For wounds with medium to high levels of exudate Stimulates wound cleansing Easy to use Sterile 4 x 4 inches, 2 x 2 inches folded
Catalog No. 50-304-5469 Supplier Med Vet International Supplier No. 400464BX
For wounds with medium to high levels of exudate Stimulates wound cleansing Easy to use Sterile 4 x 4 inches, 2 x 2 inches folded