Learn More
Med Vet International McKesson Non-Adherent Dressing, 2 x 3 Inch, 16-4292, 1200/Case
Catalog No. 503043478
Encompass_Preferred
Non-Adherent Dressing McKesson Nylon / Polyester 2 X 3 Inch Sterile Absorbent dressing pads for lightly draining wounds; can be used in combination with other wound care supplies to help protect the wound from debris and infection Nonwoven dressing pad with a non-stick formulation for easy removal; does not stick to the wound; promotes efficient healing Individually packed in an easy-to-peel package for rapid access, suitable for single use only to maintain hygiene; disposable for easy clean-up Nonadherent sterile pad is 2 x 3 inches and is individually wrapped in easy-to-peel package
Catalog No. 50-304-3478 Supplier Med Vet International Supplier No. 373768CS
Non-Adherent Dressing McKesson Nylon / Polyester 2 X 3 Inch Sterile Absorbent dressing pads for lightly draining wounds; can be used in combination with other wound care supplies to help protect the wound from debris and infection Nonwoven dressing pad with a non-stick formulation for easy removal; does not stick to the wound; promotes efficient healing Individually packed in an easy-to-peel package for rapid access, suitable for single use only to maintain hygiene; disposable for easy clean-up Nonadherent sterile pad is 2 x 3 inches and is individually wrapped in easy-to-peel package