Learn More
Med Vet International Non-Adherent Dressing, 3"X4", 100/Box
Catalog No. 502831255
Encompass_Preferred
The non-adherent pad is a unitized dressing with an absorbent inner layer of fiber that is laminated between a thin, highly porous polyethylene net facing. Designed specifically to help prevent over hydration and maceration at the wound site and surrounding skin, the pad allows for a shorter healing time. This dressing may be used with or without a secondary dressing, depending on the need for additional protection or absorption. It will not stick to the wound! Compare and Save!! These are exactly the same as top name brands with a lower price. 100 per box.
Catalog No. 50-283-1255 Supplier Med Vet International Supplier No. 9521
The non-adherent pad is a unitized dressing with an absorbent inner layer of fiber that is laminated between a thin, highly porous polyethylene net facing. Designed specifically to help prevent over hydration and maceration at the wound site and surrounding skin, the pad allows for a shorter healing time. This dressing may be used with or without a secondary dressing, depending on the need for additional protection or absorption. It will not stick to the wound! Compare and Save!! These are exactly the same as top name brands with a lower price. 100 per box.