Learn More
Med Vet International Non-Adherent Dressing, 2" X 3", 100/Box
Catalog No. 502522226
Encompass_Preferred
Unitized dressing with an absorbent inner layer of fiber laminated between a thin, highly porous polyethylene net facing. Designed specifically to help prevent over hydration and maceration at the wound site and surrounding skin, the pad allows for a shorter healing time. Can be used with or without a secondary dressing, depending on the need for additional protection or absorption. It will not stick to the wound! Compare and Save!! These are exactly the same as top name brands with a lower price.
Catalog No. 50-252-2226 Supplier Med Vet International Supplier No. NA2X3
Unitized dressing with an absorbent inner layer of fiber laminated between a thin, highly porous polyethylene net facing. Designed specifically to help prevent over hydration and maceration at the wound site and surrounding skin, the pad allows for a shorter healing time. Can be used with or without a secondary dressing, depending on the need for additional protection or absorption. It will not stick to the wound! Compare and Save!! These are exactly the same as top name brands with a lower price.