Learn More
Med Vet International McKesson Oil Emulsion Impregnated Dressing, 3 x 8 Inch, Sterile, 61-77042, Each
Catalog No. 503044982
Encompass_Preferred
Open mesh fabric prevents the pooling of fluid by draining exudate away from the wound site Provides a moist environment conducive to wound healing Nonadherent to maximize comfort during dressing change or removal
Catalog No. 50-304-4982 Supplier Med Vet International Supplier No. 488940EA
Open mesh fabric prevents the pooling of fluid by draining exudate away from the wound site Provides a moist environment conducive to wound healing Nonadherent to maximize comfort during dressing change or removal