Learn More
Med Vet International Optifoam Gentle Silicone-Faced Foam Wound Dressing, 7in. X 7in., Sacrum, Educational Packaging, 5/Box
Catalog No. 502829719
Encompass_Preferred
Optifoam Gentle Silicone-Faced sacrum Foam dressings has 5 unique layers with a superabsorbent core that absorbs exudate, converts it into gel, and locks it away. It is a highly comfortable foam dressing with gentle silicone adhesive that can be lifted and reapplied. Optifoam silicone faced wound dressing have a low-profile design and a moisture vapor transmission rate or MVTR that adjusts to the level of fluid and improves breathability. The design helps to collectively displace pressure when used with a pressure injury prevention protocol. Size: 7in. x 7in.Latex Free: Yes Quantity: 5/box
Catalog No. 50-282-9719 Supplier Med Vet International Supplier No. MSC2177EPZ
Optifoam Gentle Silicone-Faced sacrum Foam dressings has 5 unique layers with a superabsorbent core that absorbs exudate, converts it into gel, and locks it away. It is a highly comfortable foam dressing with gentle silicone adhesive that can be lifted and reapplied. Optifoam silicone faced wound dressing have a low-profile design and a moisture vapor transmission rate or MVTR that adjusts to the level of fluid and improves breathability. The design helps to collectively displace pressure when used with a pressure injury prevention protocol. Size: 7in. x 7in.Latex Free: Yes Quantity: 5/box