Learn More
Med Vet International Dynarex Sterile Dynaginate Ag Silver Calcium Alginate Rope Dressing, 1/4" X 12", 50/Case
Catalog No. 502831645
Encompass_Preferred
Calcium Alginate Dressings contours to wound bed, transforming into a soothing gel-like covering upon contact with moisture, creating an optimal moist environment with an antimicrobial barrier. Combination of alginate and silver ions that changes to a fibrous gel when in contact with wound exudate. Creates a moist wound environment and provides an antimicrobial barrier. Suitable for infected wounds or where there is a risk of infection. High absorbency dressing used for heavily exuding wounds. Primary dressing ideal for the management of partial and full thickness wounds at stages 3 or 4.
Catalog No. 50-283-1645 Supplier Med Vet International Supplier No. DYN3033
Add to Cart
Add to Cart
Calcium Alginate Dressings contours to wound bed, transforming into a soothing gel-like covering upon contact with moisture, creating an optimal moist environment with an antimicrobial barrier. Combination of alginate and silver ions that changes to a fibrous gel when in contact with wound exudate. Creates a moist wound environment and provides an antimicrobial barrier. Suitable for infected wounds or where there is a risk of infection. High absorbency dressing used for heavily exuding wounds. Primary dressing ideal for the management of partial and full thickness wounds at stages 3 or 4.