Learn More
Med Vet International Foam Dressing Allevyn Life 6-3/4 X 6-7/8 Inch With Border Film Backing Silicone Gel Adhesive Sacral Sterile, 66801306, 60/Case
Catalog No. 503043901
Encompass_Preferred
ALLEVYN LIFE is a silicone-gel adhesive, composite hydrocellular foam dressing Redistributes pressure more evenly than traditional foam dressing Up to 7 day wear time (except on the sacrum, which is up to a 5 day wear time) Indicated for shallow, granulating wounds, chronic and acute exudative wounds, full- and partial-thickness wounds such as pressure ulcers, leg ulcers, diabetic foot ulcers, infected wounds, malignant wounds, surgical wounds, first- and second-degree burns, donor sites, skin tears and fungating ulcers
Catalog No. 50-304-3901 Supplier Med Vet International Supplier No. 839950CS
ALLEVYN LIFE is a silicone-gel adhesive, composite hydrocellular foam dressing Redistributes pressure more evenly than traditional foam dressing Up to 7 day wear time (except on the sacrum, which is up to a 5 day wear time) Indicated for shallow, granulating wounds, chronic and acute exudative wounds, full- and partial-thickness wounds such as pressure ulcers, leg ulcers, diabetic foot ulcers, infected wounds, malignant wounds, surgical wounds, first- and second-degree burns, donor sites, skin tears and fungating ulcers