Learn More
Med Vet International Transparent Film Dressing With Pad 3M Tegaderm 2 X 2-3/4 inch Frame Style DelIVery Rectangle Sterile, 3582, 50/Box
Catalog No. 502831035
Encompass_Preferred
The sterile 3M Tegaderm pad creates a protective barrier to fight against infections It won't adhere to the wound bed for painless removal The pad absorbs moisture while allowing airflow Waterproof construction blocks out moisture so you can bathe or shower easily It conforms and stretches with the skin to maximize wear time The film material is transparent for easy visibility 2 x 2-3/4 inches
Catalog No. 50-283-1035 Supplier Med Vet International Supplier No. 311916BX
The sterile 3M Tegaderm pad creates a protective barrier to fight against infections It won't adhere to the wound bed for painless removal The pad absorbs moisture while allowing airflow Waterproof construction blocks out moisture so you can bathe or shower easily It conforms and stretches with the skin to maximize wear time The film material is transparent for easy visibility 2 x 2-3/4 inches