Learn More
Med Vet International Transparent Film Dressing 3M Tegaderm 4 X 4-3/4 inch Frame Style DelIVery Rectangle Sterile, 1616, 50/Box
Catalog No. 502832299
Encompass_Preferred
Provides a waterproof, sterile barrier to external contaminants including liquids, bacteria and viruses Gentle to the skin, yet holds catheters and other devices in place Hypoallergenic Conformable and flexes with skin for greater comfort
Catalog No. 50-283-2299 Supplier Med Vet International Supplier No. 468779BX
Provides a waterproof, sterile barrier to external contaminants including liquids, bacteria and viruses Gentle to the skin, yet holds catheters and other devices in place Hypoallergenic Conformable and flexes with skin for greater comfort