Learn More
Med Vet International Tegaderm Frame Style Transparent Dressing, 4in. X 4.75in., Each
Catalog No. 502829743
Encompass_Preferred
Water resistant and specially shaped to stay adherent in moist conditions. Can be cut into a variety of shapes to suit your unique needs. Stretches with the skin for best comfort. Transparent film to allow for wound monitoring in between dressing changes. Latex Free
Catalog No. 50-282-9743 Supplier Med Vet International Supplier No. MMM1626H
Water resistant and specially shaped to stay adherent in moist conditions. Can be cut into a variety of shapes to suit your unique needs. Stretches with the skin for best comfort. Transparent film to allow for wound monitoring in between dressing changes. Latex Free