Learn More
Med Vet International Transparent Film Dressing 3M Tegaderm 6 X 8 inch Frame Style DelIVery Rectangle Sterile, 1628, 10/Box
Catalog No. 502832447
Encompass_Preferred
3M™ Tegaderm™ film dressing has a frame style delivery system for better control and accurate positioning It forms a protective sterile barrier to lock out external contaminants for healing to occur Transparent construction provides high visibility for easy observation of the affected site The adhesive is latex-free and hypoallergenic to accommodate different skin types It bonds well to hold medical devices like catheters firmly in place It peels off gently for reduced pain in between dressing changes Wear time is up to 7 days
Catalog No. 50-283-2447 Supplier Med Vet International Supplier No. 121653BX
3M™ Tegaderm™ film dressing has a frame style delivery system for better control and accurate positioning It forms a protective sterile barrier to lock out external contaminants for healing to occur Transparent construction provides high visibility for easy observation of the affected site The adhesive is latex-free and hypoallergenic to accommodate different skin types It bonds well to hold medical devices like catheters firmly in place It peels off gently for reduced pain in between dressing changes Wear time is up to 7 days