Learn More
Med Vet International Transparent Film Dressing 3M Tegaderm 8 X 12 inch Frame Style DelIVery Rectangle Sterile, 1629, 10/Box
Catalog No. 502828969
Encompass_Preferred
Provides a waterproof, sterile barrier to external contaminants, including liquids, bacteria and viruses Transparent dressing allows for continuous visibility to the IV insertion site Adhesive is gentle on skin, and conformable dressing flexes with skin for comfort Picture-frame delivery makes accurate placement easy
Catalog No. 50-282-8969 Supplier Med Vet International Supplier No. 124292BX
Provides a waterproof, sterile barrier to external contaminants, including liquids, bacteria and viruses Transparent dressing allows for continuous visibility to the IV insertion site Adhesive is gentle on skin, and conformable dressing flexes with skin for comfort Picture-frame delivery makes accurate placement easy