Learn More
Med Vet International Xeroform Petrolatum Impregnated Dressing Xeroform™ Occlusive Strip 1 X 8 Inch Sterile, 8884433301, 50/Box
Catalog No. 503044424
Encompass_Preferred
Nonadherent 3% bismuth tribromophenate in a water-in-oil emulsion on fine mesh gauze For minimum to heavy exudate Clings and conforms Hydrophilic ointment Bacteriostatic action Sterile
Catalog No. 50-304-4424 Supplier Med Vet International Supplier No. 147128BX
Nonadherent 3% bismuth tribromophenate in a water-in-oil emulsion on fine mesh gauze For minimum to heavy exudate Clings and conforms Hydrophilic ointment Bacteriostatic action Sterile