Learn More
Med Vet International Xeroform Petrolatum Impregnated Dressing McKesson Strip 1 X 8 Inch Sterile, 2201, 50/Box
Catalog No. 503045243
Encompass_Preferred
McKesson Xeroform Petrolatum Dressing 1 x 8 Inch (2.5 cm x 20.3 cm) Sterile Contains 3% Bismuth Tribromophenate to provide bacteriostatic properties and help to reduce risk of infection and deodorize the wound. Primary dressing for wounds with light or no exudate. Helps maintain a moist wound healing environment to assist in autolytic debridement. Single Use Prescription Item - Federal (USA) Law Prohibits Dispensing Without A Prescription Not Made with Natural Rubber Latex
Catalog No. 50-304-5243 Supplier Med Vet International Supplier No. 864638BX
McKesson Xeroform Petrolatum Dressing 1 x 8 Inch (2.5 cm x 20.3 cm) Sterile Contains 3% Bismuth Tribromophenate to provide bacteriostatic properties and help to reduce risk of infection and deodorize the wound. Primary dressing for wounds with light or no exudate. Helps maintain a moist wound healing environment to assist in autolytic debridement. Single Use Prescription Item - Federal (USA) Law Prohibits Dispensing Without A Prescription Not Made with Natural Rubber Latex