Learn More
Med Vet International Curad Xeroform Sterile Petrolatum Gauze Dressing, 1" x 8", Each
Catalog No. 502091855
Encompass_Preferred
Petrolatum gauze with xeroform, a medicating and deodorizing agent with 3% Bismuth Tribromophenate.Occlusive non-adherent dressing contraindicated for draining wounds. Sterile, latex-free.
Catalog No. 50-209-1855 Supplier Med Vet International Supplier No. CUR253180H
Petrolatum gauze with xeroform, a medicating and deodorizing agent with 3% Bismuth Tribromophenate.Occlusive non-adherent dressing contraindicated for draining wounds. Sterile, latex-free.