Learn More
Med Vet International ComfortFoam™ Silicone Adhesive without Border Silicone Foam Dressing, 4 x 4 Inch, Sterile, 00315E, 10/Box
Catalog No. 503044158
Encompass_Preferred
Can be cut to fit wound site Silicone adhesive sticks to surrounding skin but not to the wound bed, minimizing pain and the risk of damage to the wound area Can be safely worn for several days, depending on wound condition and exudate levels
Catalog No. 50-304-4158 Supplier Med Vet International Supplier No. 835615BX
Can be cut to fit wound site Silicone adhesive sticks to surrounding skin but not to the wound bed, minimizing pain and the risk of damage to the wound area Can be safely worn for several days, depending on wound condition and exudate levels