Learn More
Med Vet International Foam Dressing Allevyn Heel 4-1/2 X 5-1/2 Inch Without Border Film Backing Nonadhesive Heel Sterile, 66007630, 5/Box
Catalog No. 503043482
Encompass_Preferred
Absorbent hydrocellular pad with a non-adherent wound contact layer and breathable top film maintains an optimal moist wound environment Breathable outer polyurethane top film helps to prevent strike-through and provides an effective barrier to bacteria Hydrocellular foam core is highly absorbent Won't stick to the wound and is suitable for use on fragile skin Cup design is made to conform to curved areas of the body, such as the heels and elbows
Catalog No. 50-304-3482 Supplier Med Vet International Supplier No. 407366BX
Absorbent hydrocellular pad with a non-adherent wound contact layer and breathable top film maintains an optimal moist wound environment Breathable outer polyurethane top film helps to prevent strike-through and provides an effective barrier to bacteria Hydrocellular foam core is highly absorbent Won't stick to the wound and is suitable for use on fragile skin Cup design is made to conform to curved areas of the body, such as the heels and elbows