Learn More
Med Vet International Foam Dressing ComfortFoam™ Border 7-1/5 X 7-1/5 Inch With Border Waterproof Backing Silicone Adhesive Sacral Sterile, 43880, 5/Box
Catalog No. 503045329
Encompass_Preferred
The 7-1/5- x 7-1/5-inch DermaRite ComfortFoam border facilitates faster and proper healing of wounds A multilayer design optimizes wound protection, thermal insulation and absorbency The skin-friendly adhesive foam dressing sticks well and removes easily A low-profile border improves adhesion and flexibility for application in difficult areas
Catalog No. 50-304-5329 Supplier Med Vet International Supplier No. 944933BX
The 7-1/5- x 7-1/5-inch DermaRite ComfortFoam border facilitates faster and proper healing of wounds A multilayer design optimizes wound protection, thermal insulation and absorbency The skin-friendly adhesive foam dressing sticks well and removes easily A low-profile border improves adhesion and flexibility for application in difficult areas