Learn More
Med Vet International DermaSyn Wound Dressing Gel, 3 oz., 00247, 24/Case
Catalog No. 503043765
Encompass_Preferred
Non-oily hydrogel wound dressing enriched with vitamin E Assists in autolytic debridement Tube for maximum coverage where hands-off application is needed Easy application and no waste
Catalog No. 50-304-3765 Supplier Med Vet International Supplier No. 584131CS
Non-oily hydrogel wound dressing enriched with vitamin E Assists in autolytic debridement Tube for maximum coverage where hands-off application is needed Easy application and no waste