Learn More
Med Vet International Super Absorbent Dressing EXU-DRY Anti-Shear 3 X 4 Inch Rectangle, 5999034, 100/Case
Supplier:  Med Vet International 330491CS
Encompass_Preferred
Exu-Dry is a one-piece wound dressing made up of multiple layers Outer Layer - High density polyethylene, Inner Layers - Highly absorbent rayon/cellulose blend, Anti Shear Layer - High density polyethylene, Wound Contact Layer - High density polyethylene Non-adherent to the wound, EXU-DRY is more comfortable for patients during and between dressing changes EXU DRY's anti-shear Layer helps reduce shearing thus may help protect fragile granulation tissue Highly absorbent, EXU DRY helps reduce the risk of maceration and irritation to the wound and surrounding tissue EXU DRY wicks away drainage from the wound into the entire dressing, reducing the frequency of dressing changes Because EXU DRY is permeable and non-occlusive, it can be used as a wet or dry dressing on clean or contaminated wounds Compatible with most topical agents, liquids and creams Full absorbency
Catalog No. 50-304-4692