Learn More
Med Vet International Dynarex Sterile Siligentle Ag Silver Silicone Bordered Foam Dressing, 4" X 4", 50/Case
Catalog No. 502831636
Encompass_Preferred
Soft and adaptable dressings, providing a moist wound environment with an antimicrobial effect and a silver compound. Superior absorption effectively locks away exudate while releasing silver for optimal healing. Soft and conformable, dressing with silver compound providing an antimicrobial barrier. Suitable for wounds with delayed healing due to bacteria, infection, or wounds at risk of re-infection. The silver foam core absorbs exudate, while releasing silver ions into the wound bed for an antibacterial effect. Vapor permeable waterproof film and silicone contact layer that is gentle on wounds and moves without losing adherence. High absorbency dressing used for heavily exuding wounds. Primary or secondary dressing to treat and manage partial to full thickness wounds at stages 2, 3 or 4.
Catalog No. 50-283-1636 Supplier Med Vet International Supplier No. 3085
Soft and adaptable dressings, providing a moist wound environment with an antimicrobial effect and a silver compound. Superior absorption effectively locks away exudate while releasing silver for optimal healing. Soft and conformable, dressing with silver compound providing an antimicrobial barrier. Suitable for wounds with delayed healing due to bacteria, infection, or wounds at risk of re-infection. The silver foam core absorbs exudate, while releasing silver ions into the wound bed for an antibacterial effect. Vapor permeable waterproof film and silicone contact layer that is gentle on wounds and moves without losing adherence. High absorbency dressing used for heavily exuding wounds. Primary or secondary dressing to treat and manage partial to full thickness wounds at stages 2, 3 or 4.