Learn More
Med Vet International Allevyn Gentle Border Heel Silicone Gel Adhesive with Border Silicone Foam Dressing, 9 x 9-1/8 Inch, Sterile, 66800506, 5/Box
Catalog No. 503045046
Encompass_Preferred
Advanced triple-layer construction with an absorbent hydrocellular pad sandwiched between a perforated silicone gel adhesive wound contact layer and a highly permeable waterproof outer film Lift and reposition the dressing without its losing its adherent properties Easy to fit and can be left in place without the need for secondary retention, conforming well to the complex contours of the heel 23 x 23.2 centimeters
Catalog No. 50-304-5046 Supplier Med Vet International Supplier No. 729695BX
Advanced triple-layer construction with an absorbent hydrocellular pad sandwiched between a perforated silicone gel adhesive wound contact layer and a highly permeable waterproof outer film Lift and reposition the dressing without its losing its adherent properties Easy to fit and can be left in place without the need for secondary retention, conforming well to the complex contours of the heel 23 x 23.2 centimeters