Learn More
Med Vet International Allevyn Gentle Border Heel Silicone Gel Adhesive with Border Silicone Foam Dressing, 9 x 9-1/8 Inch, Sterile, 66800506, 35/Case
Catalog No. 503045047
Encompass_Preferred
Advanced triple-layer construction with an absorbent hydrocellular pad sandwiched between a perforated silicone gel adhesive wound contact layer and a highly permeable waterproof outer film Lift and reposition the dressing without its losing its adherent properties Easy to fit and can be left in place without the need for secondary retention, conforming well to the complex contours of the heel 23 x 23.2 centimeters
Catalog No. 50-304-5047 Supplier Med Vet International Supplier No. 729695CS
Advanced triple-layer construction with an absorbent hydrocellular pad sandwiched between a perforated silicone gel adhesive wound contact layer and a highly permeable waterproof outer film Lift and reposition the dressing without its losing its adherent properties Easy to fit and can be left in place without the need for secondary retention, conforming well to the complex contours of the heel 23 x 23.2 centimeters