Learn More
Med Vet International Foam Dressing Aquacel 5-1/2 X 8 Inch With Border Waterproof Film Backing Silicone Adhesive Heel Sterile, 420625, 5/Box
Catalog No. 503043304
Encompass_Preferred
Hydrofiber technology absorbs wound fluid and creates a soft gel to maintain a moist wound environment Locks in exudate through vertically wicking, reducing the risk of maceration Helps minimize pain while in place and during dressing changes Silicone adhesive is gentle on skin Barrier prevents water and bacteria from getting in 5-1/2 x 8 inches for heels; 3-2/5 x 5½ inch pad
Catalog No. 50-304-3304 Supplier Med Vet International Supplier No. 802597BX
Hydrofiber technology absorbs wound fluid and creates a soft gel to maintain a moist wound environment Locks in exudate through vertically wicking, reducing the risk of maceration Helps minimize pain while in place and during dressing changes Silicone adhesive is gentle on skin Barrier prevents water and bacteria from getting in 5-1/2 x 8 inches for heels; 3-2/5 x 5½ inch pad