Learn More
Med Vet International Medline Waterproof Adhesive Stratasorb Composite Island Dressing, 4in X 10in With 2in X 8in Pad, 100/Case
Catalog No. 502831218
Encompass_Preferred
Waterproof composite dressings have four distinct layers for versatile use, including as a primary or secondary dressing Layer 1 is a nonadherent contact layer that protects the wound site and will not stick to granulating tissue Layer 2 is a soft pad that absorbs drainage and cushions Layer 3 is a nonwoven backing with an adhesive border to securely hold the dressing in place Layer 4 is a semiocclusive, waterproof polyurethane film that helps deter external contaminants and maintain an optimal wound environment
Catalog No. 50-283-1218 Supplier Med Vet International Supplier No. MSC30410
Waterproof composite dressings have four distinct layers for versatile use, including as a primary or secondary dressing Layer 1 is a nonadherent contact layer that protects the wound site and will not stick to granulating tissue Layer 2 is a soft pad that absorbs drainage and cushions Layer 3 is a nonwoven backing with an adhesive border to securely hold the dressing in place Layer 4 is a semiocclusive, waterproof polyurethane film that helps deter external contaminants and maintain an optimal wound environment