Learn More
Med Vet International Thin Foam Dressing Mepilex Transfer 8 X 20 Inch Without Border Film Backing Silicone Adhesive Rectangle Sterile, 294599, Each
Catalog No. 503044157
Encompass_Preferred
Mepilex Transfer is thin and conformable for even the most difficult to dress wounds The Safetac Technology layer allows for direct contact to the wound and the surrounding skin, including uneven surfaces May be cut to shape Thin and highly conformable Suitable for large and/or fragile wounds
Catalog No. 50-304-4157 Supplier Med Vet International Supplier No. 839941EA
Mepilex Transfer is thin and conformable for even the most difficult to dress wounds The Safetac Technology layer allows for direct contact to the wound and the surrounding skin, including uneven surfaces May be cut to shape Thin and highly conformable Suitable for large and/or fragile wounds