Learn More
Med Vet International Foam Dressing PolyMem 3 X 3 Inch Without Border Film Backing Nonadhesive Square Sterile, 5033, 15/Box
Catalog No. 503043516
Encompass_Preferred
The sterile PolyMem wound dressing is lightweight and non-adherent for quick dressing changes It's formulated with a starch co-polymer for moisture retention and includes glycerol to regulate hydration PolyMem 3-inch x 3-inch foam dressing absorbs output from the wound to keep it clean The nonadherent properties keep the wound bed protected It expands to conform to the wound, creating a protective cover
Catalog No. 50-304-3516 Supplier Med Vet International Supplier No. 258749BX
The sterile PolyMem wound dressing is lightweight and non-adherent for quick dressing changes It's formulated with a starch co-polymer for moisture retention and includes glycerol to regulate hydration PolyMem 3-inch x 3-inch foam dressing absorbs output from the wound to keep it clean The nonadherent properties keep the wound bed protected It expands to conform to the wound, creating a protective cover