Learn More
Med Vet International DermaLevin Adhesive with Border Foam Dressing, 4 x 4 Inch, Sterile, 00280E, 10/Box
Catalog No. 503042866
Encompass_Preferred
DermaLevin is a moderately absorbent waterproof foam island dressing Waterproof backing Maintains a moist wound environment Will not adhere to wound bed Conforms easily to awkward-to-dress areas
Catalog No. 50-304-2866 Supplier Med Vet International Supplier No. 896849BX
DermaLevin is a moderately absorbent waterproof foam island dressing Waterproof backing Maintains a moist wound environment Will not adhere to wound bed Conforms easily to awkward-to-dress areas