Learn More
Med Vet International McKesson Super Absorbent Dressing, 6 x 9 Inch, Sterile, 61-89569, 10/Box
Catalog No. 503043726
Encompass_Preferred
Super absorbent polymers bind drainage and convert it to a gel These dressings hold more fluid, lock fluid inside the dressing, helping to decrease the risk of maceration and keep the harmful wound fluid components away from the wound and peri-wound skin Protective outer layer and non-adherent wound contact layer for easy removal Can be used as a primary or secondary dressing for moderate to heavy exudate 6 x 9 inches
Catalog No. 50-304-3726 Supplier Med Vet International Supplier No. 682971BX
Super absorbent polymers bind drainage and convert it to a gel These dressings hold more fluid, lock fluid inside the dressing, helping to decrease the risk of maceration and keep the harmful wound fluid components away from the wound and peri-wound skin Protective outer layer and non-adherent wound contact layer for easy removal Can be used as a primary or secondary dressing for moderate to heavy exudate 6 x 9 inches