Learn More
Med Vet International Dynarex View Guard Transparent Dressings, Sterile, 4" X 4 3/4", 200/Case
Catalog No. 502830931
Encompass_Preferred
View Guard Transparent dressings protect the skin, IV sites and surgical incisions. Easy to apply Conforms to difficult body contours Provides a breathable barrier that lets air in while keeping moisture vapors and fluid out Ideal for use around IV sites, for wound care and surgical site protection
Catalog No. 50-283-0931 Supplier Med Vet International Supplier No. DYN3643
View Guard Transparent dressings protect the skin, IV sites and surgical incisions. Easy to apply Conforms to difficult body contours Provides a breathable barrier that lets air in while keeping moisture vapors and fluid out Ideal for use around IV sites, for wound care and surgical site protection