Learn More
Med Vet International Dynarex View Guard Transparent Dressings, Sterile, 6" X 8", 80/Case
Supplier:  Med Vet International DYN3644
Encompass_Preferred
View Guard Transparent dressings protect the skin, IV sites and surgical incisions. Easy to apply Conforms to difficult body contours Provides a breathable barrier that lets air in while keeping moisture vapors and fluid out Ideal for use around IV sites, for wound care and surgical site protection
Catalog No. 50-283-1555