Learn More
Medchemexpress LLC HY-125331 5mg Medchemexpress, DRF-1042 CAS:200619-13-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1253315MG
Encompass_Preferred
Medchemexpress, HY-125331 5mg DRF-1042 CAS:200619-13-2 DRF-1042 is an orally active derivative of Camptothecin. DRF-1042 acts to inhibit DNA topoisomerase I. DRF-1042 shows good anticancer activity against a panel of human cancer cell lines including multi-drug resistance (MDR) phenotype. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9691