Learn More
Aviva Systems Biology Corp DRGX Antibody - middle region (ARP47554_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47554_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DRGX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DRGX. Alternate names: DRG11, PRRXL1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59586