Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Ethyl Alcohol Drugs of Abuse Calibrators

Catalog No. 23278411 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 x 5 mL
1 x 25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-411 1 x 25 mL
23-278-316 1 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-278-411 Supplier Thermo Scientific™ Supplier No. 1406
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Choose Thermo Scientific™ DRI™ Ethyl Alcohol Drugs of Abuse Calibrators to deliver high analytical accuracy while eliminating time-consuming steps in reagent preparation.

Choose Thermo Scientific™ DRI™ Ethyl Alcohol Drugs of Abuse Calibrators to deliver high analytical accuracy while eliminating time-consuming steps in reagent preparation. Ready-to-use in liquid form, these assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description DRI Alcohol Drugs of Abuse Calibrators
Concentration 100 mg/dL
Detectable Analytes Alcohol
Sample Type Plasma, Serum, Urine
Type Calibrator
Packaging 1 Bottle
Quantity 1 x 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.