Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Ethyl Alcohol Drugs of Abuse Controls

Catalog No. 23278317 Shop All Thermo Scientific Products
Change view
Click to view available options
Concentration:
300 ng/mL
50 ng/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
23-278-317 300 ng/mL
23-278-315 50 ng/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-278-317 Supplier Thermo Scientific™ Supplier No. 0243
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eliminate time-consuming steps yet maintain high analytical accuracy with ready-to-use Thermo Scientific™ DRI™ Ethyl Alcohol Drugs of Abuse Controls in liquid form.

Eliminate time-consuming steps yet maintain high analytical accuracy with ready-to-use Thermo Scientific™ DRI™ Ethyl Alcohol Drugs of Abuse Controls in liquid form. These assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
TRUSTED_SUSTAINABILITY

Specifications

Description DRI Alcohol Drugs of Abuse Controls
Concentration 300 ng/mL
Detectable Analytes Alcohol
Sample Type Plasma, Serum, Urine
Type Control
Ethanol Ethanol
Quantity 1 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.