Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Alcohol Negative Drugs of Abuse Calibrators

Catalog No. 23278410 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 x 5 mL
1 x 25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-410 1 x 25 mL
23-278-318 1 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-278-410 Supplier Thermo Scientific™ Supplier No. 1405
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eliminate time-consuming steps in reagent preparation while still achieving high analytical accuracy.

Eliminate time-consuming steps in reagent preparation while still achieving high analytical accuracy. Ready-to-use in liquid form, Thermo Scientific™ DRI™ Alcohol Negative Drugs of Abuse Calibrators have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Do not Freeze; 2° to 8°C
Description DRI Alcohol Negative Drugs of Abuse Calibrators
Concentration 0 ng/mL
Detectable Analytes Alcohol
Sample Type Plasma, Serum, Urine
Type Calibrator
Packaging 1 Bottle
Quantity 1 x 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.