Learn More
Electron Microscopy Sciences Dri-Bath™ Digital Double Block, 120 V, 22.1 x 9 x 28.4 cm, 37°C ± 0.1 to 0.2°C Temperature
SDP
Catalog No. 5032263
Encompass_Preferred
Compact design with coated durable steel body. Temperature range ambient +5 to 130 degree C. Holds modular blocks to accommodate variety of vessels.
Dimensions: 22.1 x 9 x 28.4 cm
Temperature: 37°C ± 01 to 0.2°C
Uniformity: 37°C± 0.1 to 0.4°C
Catalog No. 50-322-63 Supplier Electron Microscopy Sciences Supplier No. 6312620
Compact design with coated durable steel body. Temperature range ambient +5 to 130 degree C. Holds modular blocks to accommodate variety of vessels.
Dimensions: 22.1 x 9 x 28.4 cm
Temperature: 37°C ± 01 to 0.2°C
Uniformity: 37°C± 0.1 to 0.4°C