Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Cocaine Drugs of Abuse Assays

Catalog No. 23278304
Change view
Click to view available options
Quantity:
3 x 18 mL
1 x 100 mL
1 x 500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-304 1 x 100 mL
21-200-408 1 x 500 mL
23-278-350 3 x 18 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-278-304 Supplier Thermo Scientific™ Supplier No. 0055
Add to Cart
Add to Cart
This item is not returnable. View return policy

Save time in reagent preparation with liquid, ready-to-use assays.

Save time in reagent preparation with liquid, ready-to-use assays. Thermo Scientific™ DRI™ Cocaine Drugs of Abuse Assays offer applications for an array of clinical chemistry analyzers. These assays have been field-tested on a wide range of analyzers to deliver high analytical accuracy and are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets MGC Multi-Drug Controls
Description DRI Cocaine Drugs of Abuse Assays
Detectable Analytes Benzoylecgonine
DoA Calibrators Multi-drug Calibrator
Sample Type Urine
Type Reagent
Quantity 1 x 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.