Learn More
Thermo Scientific™ DRI™ Cocaine Drugs of Abuse Assays
Description
Save time in reagent preparation with liquid, ready-to-use assays. Thermo Scientific™ DRI™ Cocaine Drugs of Abuse Assays offer applications for an array of clinical chemistry analyzers. These assays have been field-tested on a wide range of analyzers to deliver high analytical accuracy and are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Variety of calibrators and multiconstituent control sets
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Control Sets | MGC Multi-Drug Controls |
| Description | DRI Cocaine Drugs of Abuse Assays |
| Detectable Analytes | Benzoylecgonine |
| DoA Calibrators | DRI Multi-drug Calibrators |
| Sample Type | Urine |
| Type | Reagent |
| Quantity | 1 x 100 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.