Learn More
Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Calibrators
Catalog No. 23278320 Shop All Thermo Scientific Products
This item is not returnable. View return policy
Catalog No. 23-278-320 Supplier Thermo Scientific™ Supplier No. 0404/DEL
This item is not returnable. View return policy
Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Calibrators.
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Cotinine Drugs of Abuse Calibrators. These liquid, ready-to-use assays have been field-tested on a wide range of analyzers. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers