Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Creatinine-Detect™ Specimen Validity Test

Catalog No. 23278432 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-278-432 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-278-432 Supplier Thermo Scientific™ Supplier No. 1797
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rise to the challenge of combating the use of additives and adulterants to mask controlled substances with the Thermo Scientific™ DRI™ Creatinine-Detect™ Specimen Validity Test.

Combat the use of additives and adulterants to mask controlled substances in urine samples with Thermo Scientific™ DRI™ Creatinine-Detect™ Specimen Validity Tests. Liquid and ready-to-use, these tests can be performed on a variety of general chemistry analyzers.

  • Associated calibrators and controls offered in convenient sizes
  • Complete and automated way to test for sample integrity and adulteration
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Description DRI Creatinine-Detect Assay Specimen Validity Test
Detectable Analytes Creatinine
Sample Type Urine
Type Reagent
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.