Learn More
MSC Dri-Eaz Dehumidifier, 130 pts Water Removal, 8.8 A
Catalog No. 5000507910
Encompass
This portable, high-performance dehumidifier offers water removal even at low humidity levels, utilizing patented cross-flow technology. It includes a Bluetooth and Wi-Fi-enabled Command Hub for remote control and viewing jobsite conditions via an app and dashboard.
  • Remote monitoring and control via Bluetooth and Wi-Fi.
  • Command Hub touchscreen displays real-time drying progress and trends.
  • Advanced Crossflow Technology for maximized efficiency in low-grain conditions.
  • Fast access to humidistat, job hours, and maintenance reminders.
  • Process outlet fits 12 in. layflat and rigid ducting.
  • Sealed, large-capacity pump basin.
  • Ergonomic design with taller handle and integrated cord/hose storage.
  • Durable rotomolded housing is stackable for storage.
  • Automatic restart after a power outage.
Catalog No. 50-005-07910 Supplier MSC Supplier No. 58412339
This portable, high-performance dehumidifier offers water removal even at low humidity levels, utilizing patented cross-flow technology. It includes a Bluetooth and Wi-Fi-enabled Command Hub for remote control and viewing jobsite conditions via an app and dashboard.
  • Remote monitoring and control via Bluetooth and Wi-Fi.
  • Command Hub touchscreen displays real-time drying progress and trends.
  • Advanced Crossflow Technology for maximized efficiency in low-grain conditions.
  • Fast access to humidistat, job hours, and maintenance reminders.
  • Process outlet fits 12 in. layflat and rigid ducting.
  • Sealed, large-capacity pump basin.
  • Ergonomic design with taller handle and integrated cord/hose storage.
  • Durable rotomolded housing is stackable for storage.
  • Automatic restart after a power outage.