Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Control Sets
GSA_VA

Catalog No. 23278362
Change view
Click to view available options
Concentration:
1250 ng/mL
375 ng/mL
625 ng/mL
750 ng/mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
23-278-362 375 ng/mL
23-278-364 625 ng/mL
23-278-365 750 ng/mL
23-278-367 1250 ng/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
For Criminal Justice and Forensic use only.
Catalog No. 23-278-362 Supplier Thermo Scientific™ Supplier No. 10015934
Add to Cart
Add to Cart
This item is not returnable. View return policy
For Criminal Justice and Forensic use only.

Save time while maintaining accuracy with these liquid, ready-to-use control sets, which are for Criminal Justice and Forensic Use Only and are for sale in the U.S.A. only.

Streamline your testing process while maintaining high analytical accuracy by utilizing Thermo Scientific™ DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Control Sets along with our Thermo Scientific DRI Ethyl Glucuronide (EtG) Drugs of Abuse Assays and Calibrators. These control sets are for Criminal Justice and Forensic Use Only and are for sale in the U.S.A. only.

  • Specific detection of Ethyl Glucuronide (EtG)
  • Qualitative or semi-quantitative modes
  • Excellent correlation to LC-MS/MS
  • Applications for an array of clinical chemistry analyzers

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description DRI Ethyl Glucuronide (EtG) 375 Control. For Criminal Justice and Forensic Use Only. For sale in the U.S.A. only.
Concentration 375 ng/mL
Detectable Analytes Ethyl Glucuronide (EtG)
Sample Type Urine
Type Control
Quantity 1 x 25 mL

For sale in the U.S.A. only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.