Learn More
Thermo Scientific™ DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Assay
Shop All Thermo Scientific ProductsDescription
Achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation with Thermo Scientific™ DRI™ Ethyl Glucuronide (EtG) Drugs of Abuse Assays. Field-tested on a wide range of clinical analyzers, including the Thermo Scientific™ Indiko™ Clinical and Specialty Chemistry System, these liquid, ready-to-use assays have been widely accepted in laboratories where cost control and optimal productivity are key issues. These assays are for Criminal Justice and Forensic Use Only and are for sale in the U.S.A. only.
- Specific detection of Ethyl Glucuronide (EtG)
- Qualitative or semi-quantitative modes
- Excellent correlation to LC-MS/MS
- Applications for an array of clinical chemistry analyzers
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Control Sets | DRI Ethyl Glucuronide (EtG) Control Sets |
| Description | DRI Ethyl Glucuronide (EtG) Drugs of Abuse Assay. Criminal Justice and Forensic use only. For sale in the U.S.A. only. |
| Detectable Analytes | Ethyl Glucuronide (EtG) |
| DoA Calibrators | Ethyl Glucuronide CE Calibrator |
| Sample Type | Urine |
| Type | Reagent |
| Quantity | 1 x 500 mL |
For sale in the U.S.A. only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.